Learn More
Bellco Glass, Inc. Universal Tray,25x25cm (25mL - 2L clamps)
SDP
Catalog No. 501946156
Encompass_Preferred
Flask clamps and universal platforms for shakers. Fits onto standard platforms. Purchase individual pieces to build various configurations. Stainless Steel universal tray 25x25cm (25ML - 2L CLAMPS). MADE IN THE USA
Catalog No. 50-194-6156 Supplier Bellco Glass, Inc. Supplier No. 774417010
Flask clamps and universal platforms for shakers. Fits onto standard platforms. Purchase individual pieces to build various configurations. Stainless Steel universal tray 25x25cm (25ML - 2L CLAMPS). MADE IN THE USA