Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kuhner Shaker Inc UNIVERSAL TRAY EXU 500MM X 42

Catalog No. 502570010
Encompass

Universal Tray EXU (500mm x 420)

Catalog No. 50-257-0010 Supplier Kuhner Shaker Inc Supplier No. SMX3002
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Universal Tray EXU (500mm x 420)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.