Learn More
Mitegen Universal V1-Puck (Uni-Puck), Holds 16 Sample Bases/Pins, For Automated Sample Mounting & Cryogenic Storage, One Puck
SDP
Catalog No. 501187175
Encompass
The Universal V1-Puck (Uni-Puck) is a versatile sample pin storage and shipping container, meticulously designed for integration with many automated sample mounting systems. This puck offers robust protection for valuable cryogenic samples, holding up to 16 sample bases or pins securely in its dedicated ports. The V1-Puck ensures organized and safe sample transport and storage. Each component is identified by a unique serial number, facilitating precise sample tracking, with custom serial numbers available upon request. This design streamlines cryo-crystallography and other cryogenic workflows, enhancing efficiency and sample security. Tools specifically for handling Universal V1-Pucks are available individually, or as part of convenient Uni-puck Starter Kits. This listing is for one puck. When ordering, please provide your desired color selection and engraving of up to 8 characters, i.e. ABCD-#### in sequence starting at '1'.
Catalog No. 50-118-7175 Supplier Mitegen Supplier No. MCP111021
The Universal V1-Puck (Uni-Puck) is a versatile sample pin storage and shipping container, meticulously designed for integration with many automated sample mounting systems. This puck offers robust protection for valuable cryogenic samples, holding up to 16 sample bases or pins securely in its dedicated ports. The V1-Puck ensures organized and safe sample transport and storage. Each component is identified by a unique serial number, facilitating precise sample tracking, with custom serial numbers available upon request. This design streamlines cryo-crystallography and other cryogenic workflows, enhancing efficiency and sample security. Tools specifically for handling Universal V1-Pucks are available individually, or as part of convenient Uni-puck Starter Kits. This listing is for one puck. When ordering, please provide your desired color selection and engraving of up to 8 characters, i.e. ABCD-#### in sequence starting at '1'.