Learn More
Mitegen Universal V1-Puck (Uni-Puck) 7 Puck Set, Holds 16 Sample Bases/Pins Each, Assorted Colors, For Automated Mounting & Cryogenic Storage
SDP
Catalog No. 501940895
Encompass
The Universal V1-Puck (Uni-Puck) Set of 7 provides a versatile solution for sample pin storage and shipping, compatible with many automated sample mounting systems. This set includes seven V1-Pucks, with one puck of each color: Black, Red, Orange, Green, Blue, Purple, and Grey, aiding in visual organization. Each individual V1-Puck offers robust protection for cryogenic samples, securely holding up to 16 sample bases or pins in its dedicated ports. Consisting of two main parts, each component features a unique serial number for identification, facilitating precise sample tracking. When ordering, please provide your desired color selection and engraving of up to 8 characters, i.e. ABCD-#### in sequence starting at '1'.
Catalog No. 501940895 Supplier Mitegen Supplier No. MCP1110217
Add to Cart
Add to Cart
The Universal V1-Puck (Uni-Puck) Set of 7 provides a versatile solution for sample pin storage and shipping, compatible with many automated sample mounting systems. This set includes seven V1-Pucks, with one puck of each color: Black, Red, Orange, Green, Blue, Purple, and Grey, aiding in visual organization. Each individual V1-Puck offers robust protection for cryogenic samples, securely holding up to 16 sample bases or pins in its dedicated ports. Consisting of two main parts, each component features a unique serial number for identification, facilitating precise sample tracking. When ordering, please provide your desired color selection and engraving of up to 8 characters, i.e. ABCD-#### in sequence starting at '1'.