Learn More
AB Sciex LLC Universal Vials - 100 Pack
Catalog No. NC0604840
Encompass
For use in combination with universal caps. Vials accommodate either run buffer or sample and can also accommodate MicroVials. Vials are precision manufactured from polymethylpentene and have been tested for chemical compatibility with commonly used CE reagents. Universal vials are manufactured for use only with PA 800 Enhanced, PA 800 Plus, CESI 8000 Plus, or P/ACE MDQ Plus instruments and should not be reused.
Catalog No. NC0604840 Supplier AB Sciex LLC Supplier No. A62251
For use in combination with universal caps. Vials accommodate either run buffer or sample and can also accommodate MicroVials. Vials are precision manufactured from polymethylpentene and have been tested for chemical compatibility with commonly used CE reagents. Universal vials are manufactured for use only with PA 800 Enhanced, PA 800 Plus, CESI 8000 Plus, or P/ACE MDQ Plus instruments and should not be reused.