Learn More
Electron Microscopy Sciences Universal Vice
SDP
Catalog No. 5029569
Encompass
A single set screw loading vise for fast and easy to hold specimens for SEM. The jaws are 1" (25 mm) long, 7/16 (11 mm) wide and 5/16" (8 mm) high. 1.8"(3.1 mm) pin.
Catalog No. 50-295-69 Supplier Electron Microscopy Sciences Supplier No. 7595001
A single set screw loading vise for fast and easy to hold specimens for SEM. The jaws are 1" (25 mm) long, 7/16 (11 mm) wide and 5/16" (8 mm) high. 1.8"(3.1 mm) pin.