Learn More
Hettich Centrifuges UNIVERSAL320R,(4)BldTubes,Package
SDP
Catalog No. 502537637 Shop All Hettich Centrifuges Products
Encompass_Preferred
Hettich UNIVERSAL 320R BLOOD TUBE PACKAGE, (1) Centrifuge, (1) Swing-out Rotor, (4) Buckets (4) Adapters 13 x 75/100, (4) Adapters 16 x 75/100, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years
Catalog No. 50-253-7637 Supplier Hettich Centrifuges Supplier No. 320RBLOOD
Add to Cart
Add to Cart
Request a Quote
Hettich UNIVERSAL 320R BLOOD TUBE PACKAGE, (1) Centrifuge, (1) Swing-out Rotor, (4) Buckets (4) Adapters 13 x 75/100, (4) Adapters 16 x 75/100, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years