Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Universol- ES liquid scintillation cocktail, MP Biomedicals™
GREENER_CHOICE

Catalog No. 88248004 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 gal.
5 gal.
4 x 1 gal.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
88248004 4 x 1 gal.
88248001 1 gal.
ICN88248005 5 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 88248004 Supplier MP Biomedicals, Inc Supplier No. 0188248004
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material Universol- ES liquid scintillation cocktail
Density 0.9 to 1.0g/mL at 20°C
Quantity 4 x 1 gal.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.