Learn More
Permegear Unjacketed Franz diffusion cell, 11.28 mm orifice diameter, 5 ml receptor volume, clear glass
SDP
Supplier:  Permegear 6G010011.2805XX
Encompass
Unjacketed Franz diffusion cell, 11.28 mm orifice diameter, 5 ml receptor volume, clear glass
Catalog No. NC3723672