Learn More
MSC Unknown Aluminum Bellows Flange, 2x2 Inch, 1 ea
Catalog No. 5000792920
Encompass
This aluminum bellows mounting flange is designed for securing bellows in vacuum systems or other applications requiring a sealed, flexible connection. It features a square design with a 2 x 2 inch inside dimension, ensuring a precise fit.
  • Made from aluminum
  • Provides secure mounting for bellows
  • Features a 2 x 2 inch inside square for precise fit
  • Suitable for vacuum systems and sealed connections
  • Manufactured in the United States
Catalog No. 50-007-92920 Supplier MSC Supplier No. 63416119
This aluminum bellows mounting flange is designed for securing bellows in vacuum systems or other applications requiring a sealed, flexible connection. It features a square design with a 2 x 2 inch inside dimension, ensuring a precise fit.
  • Made from aluminum
  • Provides secure mounting for bellows
  • Features a 2 x 2 inch inside square for precise fit
  • Suitable for vacuum systems and sealed connections
  • Manufactured in the United States