Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science Unknown Compositions, Ferrous Ammonium Sulfate
GREENER_CHOICESCI_ED

Catalog No. S44611 Shop All Aldon Products
Change view
Click to view available options
Class Size:
25 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S44611 25 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S44611 Supplier Innovating Science Supplier No. IS2532
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Determine the purity of ferrous ammonium sulfate samples via Gravimetric and Spectrophotometric Analysis

  • University Level Analytical Chemistry
  • Includes enough materials for 25 groups.
  • Teacher's Manual and Student Study Guide copymaster included.
  • Kit contains:
    • 10 x Ferrous Ammonium Sulfate Samples of Various Purities, 25 g
    • Phenanthroline, 12.5 g
    • 3 x Ammonia Solution, 50 mL

Specifications

Grade University Level
Includes 10 Ferrous Ammonium Sulfate Samples of Various Purities (25 g), Phenanthroline (12.5 g), 3 Ammonia Solution (50 mL)
Class Size 25 Lab Groups
Product Type Unknown Compositions Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.