Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science Unknown Compositions, Soda Ash
GREENER_CHOICESCI_ED

Catalog No. S44610 Shop All Aldon Products
Change view
Click to view available options
Grade:
University Level
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S44610 University Level
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S44610 Supplier Innovating Science Supplier No. IS2533
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compare two analytical methods: Gravimetry and Double Acid-Base Titration in determining the purity of a range of sodium carbonate samples.

  • University Level Analytical Chemistry
  • Includes enough materials for 25 groups.
  • Teacher's Manual and Student Study Guide copymaster included.
  • Kit contains:
    • 10 x Sodium Carbonate Samples of Various Purities, 25 g
    • Calcium Chloride, 75 g
    • 2 x Hydrochloric Acid Solution, 3 M, 500 mL
    • 2 x Sodium Hydroxide Standard Solution, 2 N, 250 mL
    • 2 x Bromothymol Blue Indicator, 30 mL
    • 2 x Methyl Orange Indicator, 30 mL

Specifications

Grade University Level
Includes 10 Sodium Carbonate Samples of Various Purities (25 g), 75 g Calcium Chloride, 2 Hydrochloric Acid Solution (3 M, 500 mL), 2 Sodium Hydroxide Standard Solution (2 N, 250 mL), 2 Bromothymol Blue Indicator (30 mL), 2 Methyl Orange Indicator (30 mL)
Class Size 25 Lab Groups
Product Type Unknown Compositions Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.