Learn More
Echelon Biosciences Research Labs UNLABELED SHUTTLE PIP CARRIER 3
SDP
Supplier:  Echelon Biosciences Research Labs P9C31PK
Encompass
Unlabeled Shuttle PIP Carrier 3 has successfully delivered the following phosphoinositides into cells: PtdIns(3)P, PtdIns(4)P, PtdIns(5)P, and their fluorescent analogs. This carrier can also be found in these Shuttle PIP kits: P-9003, P-9004, P-9005. This item is non returnable
Catalog No. NC1692000