Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc 5000 g/mL in P&T Methanol, 1 mL in 2 mL Ampoule

Catalog No. 502399016 Shop All Phenomenex Products
Encompass_Preferred

Unleaded Gasoline 50% Weathered AL0-101284

Catalog No. 50-239-9016 Supplier Phenomenex Inc Supplier No. AL0101284
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Unleaded Gasoline 50% Weathered AL0-101284

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.