Learn More
Unleaded Gasoline Standard, Unweathered, 5,000 μg/mL, 1 mL
Specifications
Specifications
| Chemical Name or Material | Unleaded gasoline solution (unweathered) |
| CAS | 8006-61-9, 67-56-1 |
| CAS Max % | 99.37, 0.63 |
| Analyte or Component Names | Methanol,gasoline (regular unleaded) |
| Element | Methanol, Gasoline (regular unleaded) |
| Solvent or Matrix | Methanol |
| Quantity | 1 x 1 mL |
| Concentration or Composition (by Analyte or Components) | 5000μg/mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.