Learn More
Electron Microscopy Sciences Lyophilization Unlined Aluminum Seals, 13 mm I.D., 20 mm O.D.
SDP
Catalog No. 501898948
Encompass_Preferred
Aluminum lyophilization seals are unlined. The center disc tears out exposing the top of the stopper to the syringe, Natural color, fit mouth ID 13mm x OD 20mm.
Catalog No. 50-189-8948 Supplier Electron Microscopy Sciences Supplier No. 6032013
Aluminum lyophilization seals are unlined. The center disc tears out exposing the top of the stopper to the syringe, Natural color, fit mouth ID 13mm x OD 20mm.