Learn More
Electron Microscopy Sciences Lyophilization Unlined Aluminum Seals, 7 mm I.D., 13 mm O.D.
SDP
Catalog No. 501898947
Encompass_Preferred
Aluminum lyophilization seals are unlined. The center disc tears out exposing the top of the stopper to the syringe, Natural color, fit mouth ID 7mm x OD 13mm.
Catalog No. 50-189-8947 Supplier Electron Microscopy Sciences Supplier No. 6032007
Aluminum lyophilization seals are unlined. The center disc tears out exposing the top of the stopper to the syringe, Natural color, fit mouth ID 7mm x OD 13mm.