Learn More
MSC UNLINED COW LEATHER GLV
Catalog No. 5000768457
Encompass
These general purpose gloves are constructed from cowhide leather, featuring a cowhide leather lining for enhanced durability and comfort. Designed for various applications, they incorporate high-visibility colors for safety.
  • Cowhide leather construction
  • Cowhide leather lining
  • Medium size
  • Keystone thumb design
  • High-visibility brown, beige, and orange colors
  • ANSI/ISEA abrasion resistance level 3
  • Protects against abrasion, heat, and puncture
  • Color-coded hem for size identification
Catalog No. 50-007-68457 Supplier MSC Supplier No. 52387883
These general purpose gloves are constructed from cowhide leather, featuring a cowhide leather lining for enhanced durability and comfort. Designed for various applications, they incorporate high-visibility colors for safety.
  • Cowhide leather construction
  • Cowhide leather lining
  • Medium size
  • Keystone thumb design
  • High-visibility brown, beige, and orange colors
  • ANSI/ISEA abrasion resistance level 3
  • Protects against abrasion, heat, and puncture
  • Color-coded hem for size identification