Learn More
New England Biolabs, Inc. Lambda DNA (dam-) – 1250 µg
SDP
Catalog No. 50811750
Encompass
Duplex DNA is isolated from bacteriophage lambda (cI857ind 1 Sam 7). The bacteriophage is grown in an E. coli host which is deficient in adenine methylase (dam-). Therefore, those deoxyadenosine positions which are methylated in a wild type E. coli host are not methylated in the dam- host. This DNA gives complete digests with restriction endonucleases that are sensitive to dam methylation. Lambda DNA (dam-) is 48,502 base pairs in length.
Catalog No. 50-811-750 Supplier New England Biolabs, Inc. Supplier No. N3013L
Duplex DNA is isolated from bacteriophage lambda (cI857ind 1 Sam 7). The bacteriophage is grown in an E. coli host which is deficient in adenine methylase (dam-). Therefore, those deoxyadenosine positions which are methylated in a wild type E. coli host are not methylated in the dam- host. This DNA gives complete digests with restriction endonucleases that are sensitive to dam methylation. Lambda DNA (dam-) is 48,502 base pairs in length.