Learn More
Electron Microscopy Sciences Unmounted Cut-Off Wheel 1"D, 0.015" thick
SDP
Supplier:  Electron Microscopy Sciences 6214204
Encompass_Preferred
Made from rubber-bonded aluminum oxide grains. Excellent for cut-off work with less heat build-up. Mount on mandrels. Excellent for use with Flex Shaft Machines.  Diameter: 1"(25.4mm)
Catalog No. 50-996-824