Learn More
Electron Microscopy Sciences Unmounted Cut-Off Wheel 7/8"D, 0.030" thick
SDP
Supplier:  Electron Microscopy Sciences 6214202
Encompass_Preferred
Made from rubber-bonded aluminum oxide grains. Excellent for cut-off work with less heat build-up. Mount on mandrels. Excellent for use with Flex Shaft Machines.  Diameter: 7/8"(22.2mm)
Catalog No. 50-996-822