Learn More
Electron Microscopy Sciences Unmounted Cut-Off Wheel 1.5"D, 0.025" thick
SDP
Supplier:  Electron Microscopy Sciences 6214207
Encompass_Preferred
Made from rubber-bonded aluminum oxide grains. Excellent for cut-off work with less heat build-up. Mount on mandrels. Excellent for use with Flex Shaft Machines.  Diameter: 1-1/2"(38.1mm
Catalog No. 50-996-827