Learn More
BURKLE INC Uno sampler, V4A/PTFE, 59.06 in. (150 cm), 1 chamber 0.57 oz. (17 ml) volume
Catalog No. 5000025906
Encompass_Preferred
The single-chamber Uno target point sampler is designed for precise sampling from a specific zone or layer of bulk materials, including powders, granules, and grains. Ideal for bags, drums, silos, big bags, or piles, it enables targeted collection from a chosen depth. Samples can be analyzed individually to assess material variability or combined into a composite for accurate representation in analysis and quality control. Made from 1.4404 (V4A) stainless steel with a lightweight PTFE core, the Uno offers exceptional durability, corrosion resistance, and chemical inertness. Color-coded caps help identify samplers for different materials, preventing mix-ups. With robust construction and precision craftsmanship, this zone sampler delivers long-lasting reliability and accurate results in industrial, laboratory, and quality assurance applications.
Catalog No. 50-000-25906 Supplier BURKLE INC Supplier No. 53161150
The single-chamber Uno target point sampler is designed for precise sampling from a specific zone or layer of bulk materials, including powders, granules, and grains. Ideal for bags, drums, silos, big bags, or piles, it enables targeted collection from a chosen depth. Samples can be analyzed individually to assess material variability or combined into a composite for accurate representation in analysis and quality control. Made from 1.4404 (V4A) stainless steel with a lightweight PTFE core, the Uno offers exceptional durability, corrosion resistance, and chemical inertness. Color-coded caps help identify samplers for different materials, preventing mix-ups. With robust construction and precision craftsmanship, this zone sampler delivers long-lasting reliability and accurate results in industrial, laboratory, and quality assurance applications.