Learn More
Azenta US Inc UNO SINGLE TUBE SCANNER
Catalog No. NC3585173
Encompass
Uno Single Tube Scanner for 2D barcodes withCryoprotection, Camera-based reader for 2D barcodes on the bottom of tubes
Catalog No. NC3585173 Supplier Azenta US Inc Supplier No. ZTSUNO80
Uno Single Tube Scanner for 2D barcodes withCryoprotection, Camera-based reader for 2D barcodes on the bottom of tubes