Learn More
Cayman Chemical Unoproston 10mg
SDP
Catalog No. 502979832
Encompass_Preferred
An agonist of FP receptors (EC50 = 4.12 µM), an active metabolite of unoprostone isopropyl ester. and a derivative of PGF2α; increases inositol phosphate production in HEK293 cells expressing the human receptor; prevents constant light-induced degradation of photoreceptors in rats at 1.2, 3.6, or 6 UG/µl
Catalog No. 50-297-9832 Supplier Cayman Chemical Supplier No. 1668010 MG
An agonist of FP receptors (EC50 = 4.12 µM), an active metabolite of unoprostone isopropyl ester. and a derivative of PGF2α; increases inositol phosphate production in HEK293 cells expressing the human receptor; prevents constant light-induced degradation of photoreceptors in rats at 1.2, 3.6, or 6 UG/µl