Learn More
Bio Rad Laboratories UNOSPHERE S SUPPORT
Supplier:  Bio Rad Laboratories 1560113
Encompass
S Support, Bio-Rad; UNOsphere; Optimized to deliver speed and throughput in capture and intermediate purification steps; Strong cation exchange support for process chromatography; 100mL
Catalog No. 50-901-2603