Learn More
Bio Rad Laboratories UNOSPHERE S SUPPORT
Catalog No. 509012466
Encompass
S Support, Bio-Rad; UNOsphere; Optimized to deliver speed and throughput in capture and intermediate purification steps; Strong cation exchange support for process chromatography; 25mL
Catalog No. 50-901-2466 Supplier Bio Rad Laboratories Supplier No. 1560111
S Support, Bio-Rad; UNOsphere; Optimized to deliver speed and throughput in capture and intermediate purification steps; Strong cation exchange support for process chromatography; 25mL