Learn More
Diagnostic Biosystems UnoVue™ Mouse AP Detection Reagent
Catalog No. 502407545
Encompass_Preferred
UnoVue Mouse AP Detection Reagent is a non-biotin one-step detection reagent suitable for demonstrating antigens in formalin-fixed paraffin-embedded tissues and frozen sections. The UnoVue detection reagents may also be used with blood smears, cytosmears, and cell preparations. UnoVue detection reagents have been developed by directly labeling anti-mouse immunoglobulins with enzymes using a proprietary tandem hyperlabelling technology. This ensures consistent and reproducible immunodetection of mouse antibodies against nuclear, cytoplasmic and membrane antigens in different types of tissues. The single step UnoVue Detection Reagent enables faster staining procedures than traditional two-step methods using biotin and avidin/streptavidin conjugates, with significantly lower background. UnoVue Mouse AP Detection Reagent is suitable for use with mouse all mouse antibodies, both monoclonal and polyclonal.
Catalog No. 50-240-7545 Supplier Diagnostic Biosystems Supplier No. MUAP1000
UnoVue Mouse AP Detection Reagent is a non-biotin one-step detection reagent suitable for demonstrating antigens in formalin-fixed paraffin-embedded tissues and frozen sections. The UnoVue detection reagents may also be used with blood smears, cytosmears, and cell preparations. UnoVue detection reagents have been developed by directly labeling anti-mouse immunoglobulins with enzymes using a proprietary tandem hyperlabelling technology. This ensures consistent and reproducible immunodetection of mouse antibodies against nuclear, cytoplasmic and membrane antigens in different types of tissues. The single step UnoVue Detection Reagent enables faster staining procedures than traditional two-step methods using biotin and avidin/streptavidin conjugates, with significantly lower background. UnoVue Mouse AP Detection Reagent is suitable for use with mouse all mouse antibodies, both monoclonal and polyclonal.