Learn More
Diagnostic Biosystems UnoVue™ Mouse/Rabbit HRP Detection Reagent
Catalog No. 502407340
Encompass_Preferred
UnoVue Mouse/Rabbit HRP Detection Reagent is a non-biotin one-step detection reagent suitable for demonstrating antigens in formalin-fixed paraffin-embedded tissues and frozen sections. The UnoVue HRP Detection Reagent may also be used with blood smears, cytosmears, and cell preparations.\nUnoVue HRP detection reagents have been developed by directly labeling anti-mouse and anti-rabbit immunoglobulins with enzymes using a proprietary tandem hyperlabelling technology. This ensures consistent and reproducible immunodetection of mouse and rabbit antibodies with a single reagent. Nuclear, cytoplasmic and membrane antigens in different types of tissues can be detected readily.
Catalog No. 50-240-7340 Supplier Diagnostic Biosystems Supplier No. MRUHRP100
UnoVue Mouse/Rabbit HRP Detection Reagent is a non-biotin one-step detection reagent suitable for demonstrating antigens in formalin-fixed paraffin-embedded tissues and frozen sections. The UnoVue HRP Detection Reagent may also be used with blood smears, cytosmears, and cell preparations.\nUnoVue HRP detection reagents have been developed by directly labeling anti-mouse and anti-rabbit immunoglobulins with enzymes using a proprietary tandem hyperlabelling technology. This ensures consistent and reproducible immunodetection of mouse and rabbit antibodies with a single reagent. Nuclear, cytoplasmic and membrane antigens in different types of tissues can be detected readily.