Learn More
Diagnostic Biosystems UNOVUE MOUSE Mouse/Rabbit UnoVue™ HRP/DAB Detection System
Supplier: Diagnostic Biosystems UMR100PD
The Mouse/Rabbit UnoVue™ HRP/DAB Detection System is a non-biotin, one-step detection system suitable for demonstrating antigens in formalin-fixed paraffin-embedded tissues and cryostat sections. The UnoVue Detection System may also be used with blood smears, cytosmears, and cell preparations. The UnoVue detector kits have been developed by directly labeling anti-mouse and anti-rabbit immunoglobulins with enzymes using a proprietary tandem hyper-labelling technology. This ensures consistent and reproducible immunodetection of mouse and rabbit antibodies with a single reagent. Nuclear, cytoplasmic and membrane antigens in different types of tissues can be detected readily. The single step UnoVue Detection System enables faster staining procedures than traditional two-step methods using biotin and avidin/streptavidin conjugates, avidin/streptavidin conjugates, with significantly lower background.