Learn More
Diagnostic Biosystems UNOVUE MOUSE HRP DETECTION REA UnoVue™ Mouse HRP Detection Reagent
Catalog No. 502407516
Encompass_Preferred
UnoVue Mouse HRP Detection Reagent is suitable for use with mouse all Ig antibodies, both monoclonal and polyclonal. The reagents can be used for manual staining or with automated staining instruments and are well suited for multiplex immunohistochemical staining assays.
Catalog No. 50-240-7516 Supplier Diagnostic Biosystems Supplier No. MUHRP100
UnoVue Mouse HRP Detection Reagent is suitable for use with mouse all Ig antibodies, both monoclonal and polyclonal. The reagents can be used for manual staining or with automated staining instruments and are well suited for multiplex immunohistochemical staining assays.