Learn More
Diagnostic Biosystems UNOVUE RABBIT AP DETECTION REA UnoVue™ Rabbit AP Detection Reagent
Supplier:  Diagnostic Biosystems RUAP100
Encompass_Preferred
The UnoVue Rabbit AP Detection Reagent is suitable for use with rabbit antibodies, both monoclonal and polyclonal. The reagents can be used for manual staining or with automated staining instruments and are well suited for multiplex immunohistochemical staining assays.
Catalog No. 50-240-7487