Learn More
Description
- Made from durable 200 lb. test material
- Provides reliable protection when sending items to a crime lab for processing
Specifications
Specifications
| Product Type | Mailer Box |
| Color | White |
| Quantity | 12/Cs. |
| Dimensions (L x W x H) | 8 x 6 x 4 in. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
