Learn More
Lonza Walkersville Unproc Hu BM Fresh 3mL
Catalog No. 5000496289
Encompass
Bone marrow aspirate is collected from healthy donors and contains 80 IU/mL (nominal) porcine heparin as an anticoagulant The fresh tissue contains CD34 hematopoietic stem and progenitor cells (HSPC) and mesenchymal stem/stromal cells (MSC) CD34 HSPC have the potential to self-renew and differentiate into all the mature blood cell lineages MSC have the potential to differentiate into several lineages including bone cartilage muscle and fat Cells are collected in a transport tube Manufactured by AllCells Additional shipping charges apply for fresh human origin products
Catalog No. 50-004-96289 Supplier Lonza Walkersville Supplier No. 3M103
Bone marrow aspirate is collected from healthy donors and contains 80 IU/mL (nominal) porcine heparin as an anticoagulant The fresh tissue contains CD34 hematopoietic stem and progenitor cells (HSPC) and mesenchymal stem/stromal cells (MSC) CD34 HSPC have the potential to self-renew and differentiate into all the mature blood cell lineages MSC have the potential to differentiate into several lineages including bone cartilage muscle and fat Cells are collected in a transport tube Manufactured by AllCells Additional shipping charges apply for fresh human origin products