Learn More
Aviva Systems Biology Corp UNQ1887 Antibody - middle region (ARP52668_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52668_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against UNQ1887. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human UNQ1887. Alternate names: DKFZp586C1324, IMP2, MDHV1887, MGC126674, MGC126676, MGC90402, PRO4332, PSL4, SPPL3, UNQ1887ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-749