Learn More
BRUKER BIOSPIN UNRACKED 4" NMR-TUBES,5MM
Catalog No. NC1894604
Encompass
UNRACKED 4" NMR-TUBES WITH CODED CLOSED CAPS FOR USE WITH SAMPLEJET. Set of certified 100 tubes with coded closed caps designed for high throughput use with SampleJet on a Bruker NMR System
Catalog No. NC1894604 Supplier BRUKER BIOSPIN Supplier No. Z172599
UNRACKED 4" NMR-TUBES WITH CODED CLOSED CAPS FOR USE WITH SAMPLEJET. Set of certified 100 tubes with coded closed caps designed for high throughput use with SampleJet on a Bruker NMR System