Learn More
ERA WATERS CORP UNREG VOLATILES QC POTABLE WAT
SDP
Catalog No. NC0291928
Encompass
One 2 mL flame-sealed ampule yields in excess of 200 mL after dilution. Contains at least 60% of the analytes randomly selected from the list below at 2-50 ug/L.
Catalog No. NC0291928 Supplier ERA WATERS CORP Supplier No. 683
One 2 mL flame-sealed ampule yields in excess of 200 mL after dilution. Contains at least 60% of the analytes randomly selected from the list below at 2-50 ug/L.