Learn More
Chiron Unsaturated Telomeric Acid, FTUCAs, and Unsaturated Acid Standards
Shop All Spex CertiPrep Products

Description
- Various singles available
- Solutions in assorted solvents
- Useful for various matrices
- Custom standards of other PFAS compounds available on demand
Specifications
Specifications
| Chemical Name or Material | 2H-Perfluoro-2-octenoic acid |
| CAS | 70887-88-6 |
| Concentration | 50 μg/mL |
| Synonym | 6:2 FTUCA |
| Type | Reference Material |
| Solvent or Matrix | Methanol |
| Quantity | 1 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.