Learn More
Cayman Chemical Robinetin, Purity: ≥98%, CAS: 490-31-3, 25 mg
SDP
Catalog No. 502913491
Encompass_Preferred
A flavonol with diverse biological activities; inhibits NADH oxidase (IC50 = 19 nmol/mg of protein in isolated beef heart mitochondria); scavenges DPPH radicals in a cell-free assay; inhibits GST (IC50 = 1.39 µM for the equine liver enzyme); inhibits MRP1 and MRP2 in MDCK-II cells (IC50s = 13.6 and 15 µM, respectively, for the human proteins)
Catalog No. 50-291-3491 Supplier Cayman Chemical Supplier No. 3527225 MG
A flavonol with diverse biological activities; inhibits NADH oxidase (IC50 = 19 nmol/mg of protein in isolated beef heart mitochondria); scavenges DPPH radicals in a cell-free assay; inhibits GST (IC50 = 1.39 µM for the equine liver enzyme); inhibits MRP1 and MRP2 in MDCK-II cells (IC50s = 13.6 and 15 µM, respectively, for the human proteins)