Learn More
New England Biolabs, Inc. Unstained Protein Standard, Broad Range – 150 gel lanes
SDP
Catalog No. 501959361
Encompass
The Unstained Protein Standard, Broad Range is a mixture of highly pure, recombinant proteins that resolves into 13 sharp bands from 10-200 kDa when electrophoresed and stained with Coomassie Brilliant Blue R-250 or other commercially available, ready-to-use protein stains.
Catalog No. 50-195-9361 Supplier New England Biolabs, Inc. Supplier No. P7717S
The Unstained Protein Standard, Broad Range is a mixture of highly pure, recombinant proteins that resolves into 13 sharp bands from 10-200 kDa when electrophoresed and stained with Coomassie Brilliant Blue R-250 or other commercially available, ready-to-use protein stains.