Learn More
Abcam Unstained Protein Ladder (10 - 200 kDa), 500UL
Catalog No. 502657326
Encompass_Preferred
Unstained Protein Ladder ab234618 contains 12 unstained recombinant proteins covering a wide range of molecular weights from 10-200 kDa.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-7326 Supplier Abcam Supplier No. AB234618500UL
Unstained Protein Ladder ab234618 contains 12 unstained recombinant proteins covering a wide range of molecular weights from 10-200 kDa.
The product is subject to the following: Abcam Restricted Use Statement