Learn More
INFORS USA INC UNV MULTITRON TRAY
SDP
Catalog No. NC0892515
Encompass
  • Universal tray design for versatile applications
  • Compatible with INFORS HT Multitron incubator shakers
  • Constructed from anodized aluminum for durability
  • Autoclavable for maintaining a sterile environment
  • Continuous grid design allows for flexible configurations
  • Supports various clamps, test tube holders, microtiter plates, or Sticky Stuff
  • Maximum capacity for 5000 mL flasks: 6 flasks
  • Maximum capacity for 25 mL flasks: 118 flasks
  • Platform dimensions: 840 x 450 mm
  • Removable design for easy cleaning
  • Engineered to support heavy loads
Catalog No. NC0892515 Supplier INFORS USA INC Supplier No. I20000
  • Universal tray design for versatile applications
  • Compatible with INFORS HT Multitron incubator shakers
  • Constructed from anodized aluminum for durability
  • Autoclavable for maintaining a sterile environment
  • Continuous grid design allows for flexible configurations
  • Supports various clamps, test tube holders, microtiter plates, or Sticky Stuff
  • Maximum capacity for 5000 mL flasks: 6 flasks
  • Maximum capacity for 25 mL flasks: 118 flasks
  • Platform dimensions: 840 x 450 mm
  • Removable design for easy cleaning
  • Engineered to support heavy loads