Learn More
Sigma Aldrich Fine Chemicals Biosciences UnwireTM full racks polyke1EA
Catalog No. 502900034
Encompass
Molded of durable polyketone. Unwire test tube racks have no coating that can chip or peel and permit corrosion. Excellent replacement for wire racks withstands most lab chemicals. Excellent thermal properties and wont get brittle in freezers. Grid design allows tubes to be tipped from side to side for clearly marking position. These racks will not float in water baths.
Catalog No. 50-290-0034 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. R83911EA
Molded of durable polyketone. Unwire test tube racks have no coating that can chip or peel and permit corrosion. Excellent replacement for wire racks withstands most lab chemicals. Excellent thermal properties and wont get brittle in freezers. Grid design allows tubes to be tipped from side to side for clearly marking position. These racks will not float in water baths.