Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Up in Vapor! Hydrogen Peroxide Decomposition Demo Kit
GREENER_CHOICESCI_ED

Catalog No. S37754 Shop All Aldon Products
Change view
Click to view available options
Grade:
7 to 14
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S37754 7 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
DOT Info: UN2014, Hydrogen peroxide, aqueous solutions, 5.1, (8), PG II UPS HAZARD CHARGE APPLIES
Catalog No. S37754 Supplier Innovating Science™ Supplier No. IS7033
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
DOT Info: UN2014, Hydrogen peroxide, aqueous solutions, 5.1, (8), PG II UPS HAZARD CHARGE APPLIES

Includes

1 x 350 mL Hydrogen Peroxide (30%) and 7 x 4 g Sodium Iodide

Shows decomposition of chemical hydrogen peroxide using sodium iodide.

Shows decomposition of chemical hydrogen peroxide using sodium iodide.

Specifications

Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Shows decomposition of chemical hydrogen peroxide using sodium iodide.
Includes 1 x 350 mL Hydrogen Peroxide (30%) and 7 x 4 g Sodium Iodide
Quantity 1
Dimensions (L x W x H) 9.75 x 7.125 x 12.25 in.
Class Size 1 Lab Group
Product Type Up in Vapor! Hydrogen Peroxide Decomposition Demo Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.