Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Upacicalcet sodium | 2052969-18-1 | 99.9% | 373.75 | 1 ML
SDP

Catalog No. 5000345224
Encompass_Preferred

Upacicalcet sodium | 2052969-18-1 | 99.9% | 373.75 | 1 ML

Catalog No. 50-003-45224 Supplier Medchemexpress LLC Supplier No. HY109106A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Upacicalcet sodium | 2052969-18-1 | 99.9% | 373.75 | 1 ML

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.