Learn More
Medchemexpress LLC Upacicalcet sodium | 2052969-18-1 | 99.9% | 373.75 g/mol | C11H13ClN3NaO6S | 5 MG
SDP
Catalog No. 5000264065
Encompass_Preferred
Upacicalcet sodium is a non-peptide calcimimetic that acts as a calcium-sensing receptor (CaSR) agonist and is used in preclinical research to reduce intact parathyroid hormone (iPTH) and serum calcium levels, with reported effects on vascular calcification and bone abnormalities in secondary hyperparathyroidism models.
  • Potent CaSR agonist (reported EC50 = 10.8 nM).
  • Reduces serum intact parathyroid hormone and lowers serum calcium.
  • Shown to improve vascular calcification and bone disorder outcomes in preclinical models.
  • Offered in multiple research quantities and as ready-to-use DMSO solutions.
  • Suitable for studies of secondary hyperparathyroidism and chronic kidney disease-related bone effects.
Catalog No. 50-002-64065 Supplier Medchemexpress LLC Supplier No. HY109106A5MG
Upacicalcet sodium is a non-peptide calcimimetic that acts as a calcium-sensing receptor (CaSR) agonist and is used in preclinical research to reduce intact parathyroid hormone (iPTH) and serum calcium levels, with reported effects on vascular calcification and bone abnormalities in secondary hyperparathyroidism models.
  • Potent CaSR agonist (reported EC50 = 10.8 nM).
  • Reduces serum intact parathyroid hormone and lowers serum calcium.
  • Shown to improve vascular calcification and bone disorder outcomes in preclinical models.
  • Offered in multiple research quantities and as ready-to-use DMSO solutions.
  • Suitable for studies of secondary hyperparathyroidism and chronic kidney disease-related bone effects.