Learn More
Medchemexpress LLC HY-19569 25mg , Upadacitinib ABT-494 CAS:1310726-60-3 Purity:98%
SDP
Catalog No. 501493985
Encompass_Preferred
Medchemexpress, HY-19569 25mg Upadacitinib ABT-494 CAS:1310726-60-3 Formula:C17H19F3N6O IC50: 43 nM (JAK1), 200 nM (JAK2) Purity:98% Upadacitinib (ABT-494) is a potent and selective Janus kinase ( JAK ) 1 inhibitor being developed for the treatment of several autoimmune disorders with an IC 50 of 43 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3985 Supplier Medchemexpress LLC Supplier No. HY1956925MG
Medchemexpress, HY-19569 25mg Upadacitinib ABT-494 CAS:1310726-60-3 Formula:C17H19F3N6O IC50: 43 nM (JAK1), 200 nM (JAK2) Purity:98% Upadacitinib (ABT-494) is a potent and selective Janus kinase ( JAK ) 1 inhibitor being developed for the treatment of several autoimmune disorders with an IC 50 of 43 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.