Learn More
Medchemexpress LLC HY-19569 10mg Medchemexpress, Upadacitinib CAS:1310726-60-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1956910MG
Encompass_Preferred
Medchemexpress, HY-19569 10mg Upadacitinib CAS:1310726-60-3 Upadacitinib (ABT-494) is a potent, orally active and selective Janus kinase 1 (JAK1) inhibitor (IC50=43 nM). Upadacitinib (ABT-494) displays approximately 74 fold selective for JAK1 over JAK2 (200 nM) in cellular assays dependent on specific, relevant cytokines. Upadacitinib (ABT-494) is used in development for the treatment of several autoimmune disorders[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9282