Learn More
TARGETMOL CHEMICALS INC UPADACITINIB 10MG
SDP
Catalog No. 502576311
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Upadacitinib (ABT-494) (ABT-494) is a selective Janus kinase (JAK) 1 inhibitor which is being studied for the treatment of several autoimmune disorders in the IC50 of 43 nM. purity: 99%
Catalog No. 50-257-6311 Supplier TARGETMOL CHEMICALS INC Supplier No. T7503
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Upadacitinib (ABT-494) (ABT-494) is a selective Janus kinase (JAK) 1 inhibitor which is being studied for the treatment of several autoimmune disorders in the IC50 of 43 nM. purity: 99%