Learn More
Medchemexpress LLC Upadacitinib-15N,d2 | 100.0% | 383.37 | C17H17D2F3N515NO | 1mg
SDP
Catalog No. 5000268632
Encompass_Preferred
Upadacitinib-15N d2 (ABT-494-15N d2) is the deuterium-labeled Upadacitinib (HY-19569) Upadacitinib-15N d2 (ABT-494) is a potent orally active and selective Janus kinase 1 (JAK1) inhibitor (IC50 43 nM) Upadacitinib-15N d2 (ABT-494) displays approximately 74 fold selective for JAK1 over JAK2 (200 nM) in cellular assays dependent on specific relevant cytokines Upadacitinib-15N d2 (ABT-494) can be used for several autoimmune disorders research[1][2]
Catalog No. 50-002-68632 Supplier Medchemexpress LLC Supplier No. HYW7278791MG
Upadacitinib-15N d2 (ABT-494-15N d2) is the deuterium-labeled Upadacitinib (HY-19569) Upadacitinib-15N d2 (ABT-494) is a potent orally active and selective Janus kinase 1 (JAK1) inhibitor (IC50 43 nM) Upadacitinib-15N d2 (ABT-494) displays approximately 74 fold selective for JAK1 over JAK2 (200 nM) in cellular assays dependent on specific relevant cytokines Upadacitinib-15N d2 (ABT-494) can be used for several autoimmune disorders research[1][2]